Läs mer
4-Methoxy-3-methylphenol, TRC
Produktkod. 30473545
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30473545 100 mg
30473547 10 mg
30466662 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30473545 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM336293100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14786-82-4
Molekylformel C8H10O2
Molekylvikt (g/mol) 138.16
IUPAC-namn 4-methoxy-3-methylphenol
LEDER COc1ccc(O)cc1C
Specifikationer
Kemiskt namn eller material 4-Methoxy-3-methylphenol
Kvantitet 100 mg
InChI formel InChI=1S/C8H10O2/c1-6-5-7(9)3-4-8(6)10-2/h3-5,9H,1-2H3
Formel vikt 138.0681