missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-methylphenylboronic acid, 97%, Thermo Scientific™

Catalog Number p-4228773
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
175883-62-2
C8H11BO3
165.98
MFCD02179464
PXVDQGVAZBTFIB-UHFFFAOYSA-N
4-methoxy-3-methylbenzeneboronic acid, 4-methoxy-3-methylphenyl boronic acid, 4-methoxy-3-methylphenyl boranediol, 3-methyl-4-methoxyphenylboronic acid, boronic acid, 4-methoxy-3-methylphenyl, boronic acid,b-4-methoxy-3-methylphenyl, b-4-methoxy-3-methylphenyl boronic acid, akos brn-0220, 4-methoxy-3-methyl-phenyl boronic acid, timtec-bb sbb005903
2773487
(4-methoxy-3-methylphenyl)boronic acid
COC1=CC=C(C=C1C)B(O)O
This item is not returnable. View return policy
175883-62-2
C8H11BO3
165.98
MFCD02179464
PXVDQGVAZBTFIB-UHFFFAOYSA-N
4-methoxy-3-methylbenzeneboronic acid, 4-methoxy-3-methylphenyl boronic acid, 4-methoxy-3-methylphenyl boranediol, 3-methyl-4-methoxyphenylboronic acid, boronic acid, 4-methoxy-3-methylphenyl, boronic acid,b-4-methoxy-3-methylphenyl, b-4-methoxy-3-methylphenyl boronic acid, akos brn-0220, 4-methoxy-3-methyl-phenyl boronic acid, timtec-bb sbb005903
2773487
(4-methoxy-3-methylphenyl)boronic acid
COC1=CC=C(C=C1C)B(O)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 219.3°C to 220.8°C
Color Purple to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 165.99
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Methoxy-3-methylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxy-3-methylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'