missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-4'-nitrobenzophenone, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Methoxy-4'-nitrobenzophenone is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxy-4′-nitrobenzophenone is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
4-Methoxy-4′-nitrobenzophenone is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 125°C to 127°C |
Quantity | 1 g |
Formula Weight | 257.25 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methoxy-4'-nitrobenzophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'