missing translation for 'onlineSavingsMsg'
Läs mer

4-Methoxy-d3-benzoic Acid, TRC

Produktkod. 30341806
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30341806 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30341806 Supplier Toronto Research Chemicals Supplier No. TRCA67330310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-Methoxybenzoic Acid D3 (methoxy-D3)
CAS 27914-54-1
Molekylformel C8H5D3O3
Kvantitet 10 mg
Synonym p-Anisic-α,α,α-d3 Acid, 4-(Trideuteromethoxy)benzoic Acid
InChI formel InChI=1S/C8H8O3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H,9,10)/i1D3
LEDER O=C(O)C1=CC=C(OC([2H])([2H])[2H])C=C1
IUPAC-namn 4-(methoxy-d3)benzoic acid
Molekylvikt (g/mol) 155.17
Formel vikt 155.07
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.