Läs mer
4-Methoxy Estrone, TRC
Produktkod. 17932905
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17932905 1 mg
17903505 5 mg
17934455 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17932905 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2261351MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 58562-33-7
Molekylformel C19 H24 O3
Molekylvikt (g/mol) 300.39
Synonym Estra-1,3,5(10)-trien-17-one, 3-hydroxy-4-methoxy-, Estrone, 4-methoxy- (6CI), 3-Hydroxy-4-methoxyestra-1,3,5(10)-trien-17-one, 4-Methoxyestrone
IUPAC-namn (8R,9S,13S,14S)-3-hydroxy-4-methoxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one
LEDER COc1c(O)ccc2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CCc12
Specifikationer
Kemiskt namn eller material 4-Methoxy Estrone
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C19H24O3/c1-19-10-9-12-11-5-7-16(20)18(22-2)14(11)4-3-13(12)15(19)6-8-17(19)21/h5,7,12-13,15,20H,3-4,6,8-10H2,1-2H3/t12-,13-,15+,19+/m1/s1
Formel vikt 300.1725
Procent renhet >95%