Läs mer
4-Methoxy Moxonidine, TRC
Produktkod. 30040633
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040633 100 mg
18021550 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30040633 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM264545100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75439-01-9
Molekylformel C10 H15 N5 O2
Molekylvikt (g/mol) 237.26
Synonym N-(Imidazolidin-2-ylidene)-4,6-dimethoxy-2-methylpyrimidin-5-amine, Moxonidine Imp. B (EP), 4-Methoxymoxonidine
IUPAC-namn N-(4,6-dimethoxy-2-methylpyrimidin-5-yl)imidazolidin-2-imine
LEDER COc1nc(C)nc(OC)c1N=C2NCCN2
Specifikationer
Kemiskt namn eller material N-(Imidazolidin-2-ylidene)-4,6-dimethoxy-2-methylpyrimidin-5-amine (4-Methoxymoxonidine)
Kvantitet 100 mg
InChI formel InChI=1S/C10H15N5O2/c1-6-13-8(16-2)7(9(14-6)17-3)15-10-11-4-5-12-10/h4-5H2,1-3H3,(H2,11,12,15)
Formel vikt 237.1226