Learn More
4-Methoxy-N-methylbenzylamine, 95%, Thermo Scientific Chemicals
Catalog Number p-4813134
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
702-24-9
C9H13NO
151.21
AIJFPNKGGAPZFJ-UHFFFAOYSA-N
4-methoxy-n-methylbenzylamine, n-4-methoxybenzyl-n-methylamine, n-methyl-4-methoxybenzylamine, 1-4-methoxyphenyl-n-methylmethanamine, 4-methoxyphenyl methyl methyl amine, benzenemethanamine, 4-methoxy-n-methyl, 4-methoxy-benzyl-methyl-amine, 4-methoxyphenyl-n-methylmethanamine, n-methyl-p-methoxybenzylamine, 1-4-methoxyphenyl-n-methyl-methanamine
485407
1-(4-methoxyphenyl)-N-methylmethanamine
CNCC1=CC=C(C=C1)OC
702-24-9
C9H13NO
151.21
AIJFPNKGGAPZFJ-UHFFFAOYSA-N
4-methoxy-n-methylbenzylamine, n-4-methoxybenzyl-n-methylamine, n-methyl-4-methoxybenzylamine, 1-4-methoxyphenyl-n-methylmethanamine, 4-methoxyphenyl methyl methyl amine, benzenemethanamine, 4-methoxy-n-methyl, 4-methoxy-benzyl-methyl-amine, 4-methoxyphenyl-n-methylmethanamine, n-methyl-p-methoxybenzylamine, 1-4-methoxyphenyl-n-methyl-methanamine
485407
1-(4-methoxyphenyl)-N-methylmethanamine
CNCC1=CC=C(C=C1)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.0080g/mL
Boiling Point 222.0°C
Flash Point 109°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2NCH3
Quantity 1 g
Specific Gravity 1.008
Solubility Information Solubility in water: slightly soluble
Formula Weight 151.21
Percent Purity 95%
Chemical Name or Material 4-Methoxy-N-methylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-(4-Methoxybenzyl)-N-methylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.