Läs mer
4-Methoxy Retinoic Acid, TRC
Produktkod. 30477041
Change view
Kvantitet:
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30477041 10 mg -
30477987 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30477041 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26780010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81121-20-2
Molekylformel C21 H30 O3
Molekylvikt (g/mol) 330.46
Synonym 4-Methoxyretinoic acid (9CI), (±)-4-Methoxyretinoic acid
IUPAC-namn (2E,4E,6E,8E)-9-(3-methoxy-2,6,6-trimethylcyclohexen-1-yl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
LEDER COC1CCC(C)(C)C(=C1C)\C=C\C(=C\C=C\C(=C\C(=O)O)\C)\C
Specifikationer
Kemiskt namn eller material 4-Methoxy Retinoic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C21H30O3/c1-15(8-7-9-16(2)14-20(22)23)10-11-18-17(3)19(24-6)12-13-21(18,4)5/h7-11,14,19H,12-13H2,1-6H3,(H,22,23)/b9-7+,11-10+,15-8+,16-14+
Formel vikt 330.2195