missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-4509800
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
2393-23-9
C8H11NO
137.18
IDPURXSQCKYKIJ-UHFFFAOYSA-N
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
75452
CHEBI:49837
(4-methoxyphenyl)methanamine
COC1=CC=C(C=C1)CN
2393-23-9
C8H11NO
137.18
IDPURXSQCKYKIJ-UHFFFAOYSA-N
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
75452
CHEBI:49837
(4-methoxyphenyl)methanamine
COC1=CC=C(C=C1)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 236.0°C to 237.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CH2NH2
Refractive Index 1.5460 to 1.5480
Quantity 25 mL
Beilstein 13,606
Specific Gravity 1.05
Formula Weight 137.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'