Läs mer
4-methoxybutan-2-ol, TRC
Produktkod. 30467266
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30467266 50 mg
30478063 100 mg
30472590 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30467266 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26640350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41223-27-2
Molekylformel C5H12O2
Molekylvikt (g/mol) 104.15
IUPAC-namn 4-methoxybutan-2-ol
LEDER COCCC(C)O
Specifikationer
Kemiskt namn eller material 4-methoxybutan-2-ol
Kvantitet 50 mg
InChI formel InChI=1S/C5H12O2/c1-5(6)3-4-7-2/h5-6H,3-4H2,1-2H3
Formel vikt 104.0837