Läs mer
4-Methoxycinnamic Acid Ethyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-Methoxycinnamic Acid Ethyl Ester |
| CAS | 1929-30-2 |
| Molekylformel | C12H14O3 |
| Kvantitet | 250 mg |
| Synonym | Ethyl 3-(4-Methoxyphenyl)-2-propenoate, Ethyl 3-(4-methoxyphenyl)acrylate, Ethyl 3-(4-Methoxyphenyl)propenoate, Ethyl 4-Methoxycinnamate, Ethyl p-Methoxycinnamate, Methyl-p-coumaric Acid Ethyl Ester, NSC 26462, NSC 44831, NSC 636698, 3-(4-Methoxyphenyl)-2-propenoic Acid Ethyl Ester, 3-(4-Methoxyphenyl)acrylic Acid Ethyl Ester, p-Methoxycinnamic Acid Ethyl Ester, 3-(4-Methoxyphenyl)-2-propenoic Acid Ethyl Ester |
| InChI formel | InChI=1S/C12H14O3/c1-3-15-12(13)9-6-10-4-7-11(14-2)8-5-10/h4-9H,3H2,1-2H3 |
| LEDER | CCOC(=O)C=Cc1ccc(OC)cc1 |
| IUPAC-namn | ethyl 3-(4-methoxyphenyl)prop-2-enoate |
| Molekylvikt (g/mol) | 206.2 |
| Formel vikt | 206.0943 |