missing translation for 'onlineSavingsMsg'
Läs mer

4-(Methoxymethyl)benzoic acid, TRC

Produktkod. 30476260
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476260 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30476260 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32983525MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 4-(Methoxymethyl)benzoic acid
CAS 67003-50-3
Molekylformel C9 H10 O3
Kvantitet 25 mg
InChI formel InChI=1S/C9H10O3/c1-12-6-7-2-4-8(5-3-7)9(10)11/h2-5H,6H2,1H3,(H,10,11)
LEDER COCc1ccc(cc1)C(=O)O
IUPAC-namn 4-(methoxymethyl)benzoic acid
Molekylvikt (g/mol) 166.174
Formel vikt 166.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.