Läs mer
4-Methoxymethylcyclohexylmethanol, TRC
Produktkod. 30473282
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30473282 250 mg
30470946 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30473282 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM262245250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Methoxymethylcyclohexylmethanol
CAS 98955-27-2
Molekylformel C9 H18 O2
Kvantitet 250 mg
Synonym 4-(Methoxymethyl)-cyclohexanemethanol
InChI formel InChI=1S/C9H18O2/c1-11-7-9-4-2-8(6-10)3-5-9/h8-10H,2-7H2,1H3
LEDER COCC1CCC(CO)CC1
IUPAC-namn [4-(methoxymethyl)cyclohexyl]methanol
Molekylvikt (g/mol) 158.238
Formel vikt 158.1307