missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
CAS Min % | 97.0 |
Density | 1.1900g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CH3OC6H4NCS |
Quantity | 10 g |
Formula Weight | 165.22 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methoxyphenyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'