Learn More
4-Methoxypyridine, 98+%
Product Code. 11385405
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11385405 25 g 25g
11375405 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11385405 Supplier Thermo Scientific Alfa Aesar Supplier No. L19625.14
 Restricted Item
I lager
Add to basket
CAS: 620-08-6 | C6H7NO | 109.128 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 620-08-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00674049
InChI Key XQABVLBGNWBWIV-UHFFFAOYSA-N
Synonym pyridine, 4-methoxy, methyl pyridin-4-yl ether, 4-methoxy pyridine, .gamma.-methoxypyridine, 4-methoxypyridin, 4-methoxypyrdine, 4-methoxylpyridine, 4-methoxy-pyridine, 4-methoxypyridine, pubchem2585
PubChem CID 69278
IUPAC Name 4-methoxypyridine
SMILES COC1=CC=NC=C1
Specifications
Melting Point 4°C
Density 1.075
Boiling Point 191°C
Flash Point 74°C (165°F)
Refractive Index 1.521
Quantity 25 g
Beilstein 108211
Formula Weight 109.13
Percent Purity ≥98%
Chemical Name or Material 4-Methoxypyridine