Läs mer
4-Methoxypyridine N-Oxide Hydrate, TRC
Produktkod. 30472203
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30472203 5 g
1 options
Produktkod. 30472203 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2671305G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Methoxypyridine N-Oxide Hydrate
CAS 304671-95-2
Molekylformel C6H9NO3
Kvantitet 5 g
Synonym 4-Methoxypyridine 1-Oxide Hydrate, NSC 27964 Hydrate, p-Methoxypyridine N-Oxide Hydrate
InChI formel InChI=1S/C6H7NO2.H2O/c1-9-6-2-4-7(8)5-3-6;/h2-5H,1H3;1H2
LEDER COc1cc[n+]([O-])cc1
Molekylvikt (g/mol) 143.14