missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxystyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.
Solubility
Slightly miscible with methanol.
Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.
Solubility
Slightly miscible with methanol.
Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
Specifications
Density | 1.002 |
Boiling Point | 204°C to 205°C |
Flash Point | 72°C (161°F) |
Refractive Index | 1.561 |
Quantity | 5 g |
Beilstein | 1098935 |
Solubility Information | Slightly miscible with methanol. |
Formula Weight | 134.18 |
Percent Purity | 98% |
Chemical Name or Material | 4-Methoxystyrene, stabilized with 0.1% 4-tert-butylcatechol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'