missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-oxazole-5-carboxylic acid, 95%, Thermo Scientific™

Catalog Number p-4510008
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
2510-32-9
C5H5NO3
127.099
ZIXUNDOOBLSXPE-UHFFFAOYSA-N
4-methyloxazole-5-carboxylic acid, 4-methyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 4-methyl, 4-methyl-oxazole-5-carboxylic acid, ksc205g3h, 4-methyl-5-oxazolecarboxylicacid, 4-methyl-oxazol-5-carboxylic acid, 4-methoxyoxazole-5-carboxylic acid, 5-oxazolecarboxylicacid, 4-methyl
292311
4-methyl-1,3-oxazole-5-carboxylic acid
CC1=C(OC=N1)C(=O)O
This item is not returnable. View return policy
2510-32-9
C5H5NO3
127.099
ZIXUNDOOBLSXPE-UHFFFAOYSA-N
4-methyloxazole-5-carboxylic acid, 4-methyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 4-methyl, 4-methyl-oxazole-5-carboxylic acid, ksc205g3h, 4-methyl-5-oxazolecarboxylicacid, 4-methyl-oxazol-5-carboxylic acid, 4-methoxyoxazole-5-carboxylic acid, 5-oxazolecarboxylicacid, 4-methyl
292311
4-methyl-1,3-oxazole-5-carboxylic acid
CC1=C(OC=N1)C(=O)O
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 127.1
Percent Purity 95%
Chemical Name or Material 4-Methyl-1, 3-oxazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'