Läs mer
(4-Methyl-1H-imidazol-5-yl)methanol Hydrochloride, TRC
Produktkod. 30482462
Change view
Kvantitet:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30482462 500 mg -
30466903 2.5 g 2.5g
30480494 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30482462 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM313575500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38585-62-5
Molekylformel C5 H8 N2 O . Cl H
Molekylvikt (g/mol) 148.59
Synonym 1H-Imidazole-5-methanol, 4-methyl-, hydrochloride (1:1), 1H-Imidazole-4-methanol, 5-methyl-, monohydrochloride (9CI), (4-Methyl-1H-imidazol-5-yl)methanol hydrochloride, (5-Methyl-1H-imidazol-4-yl)methanol hydrochloride, 4-(Hydroxymethyl)-5-methylimidazole hydrochloride, 4-Methyl-1H-imidazole-5-methanol hydrochloride, 4-Methyl-5-imidazolemethanol hydrochloride, 5-Methyl-4-imidazolemethanol hydrochloride
IUPAC-namn (5-methyl-1H-imidazol-4-yl)methanol;hydrochloride
LEDER Cl.Cc1[nH]cnc1CO
Specifikationer
Kemiskt namn eller material (5-Methyl-1H-imidazol-4-yl)methanol Hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C5H8N2O.ClH/c1-4-5(2-8)7-3-6-4;/h3,8H,2H2,1H3,(H,6,7);1H
Formel vikt 148.0403