missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(2-pyrazinyl)-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2701979
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
216959-92-1
C9H7N3O2S
221.23
MFCD00111662
XDZXQDHRHGIPRN-UHFFFAOYSA-N
4-methyl-2-2-pyrazinyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 2-5-carboxy-4-methyl-1,3-thiazol-2-yl pyrazine, 4-methyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-methyl-2-2-pyrazinyl-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl thiazole-5-carboxylic acid, 5-carboxy-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 5-thiazolecarboxylicacid, 4-methyl-2-2-pyrazinyl, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyrazinyl
2776505
4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
CC1=C(SC(=N1)C1=NC=CN=C1)C(O)=O
This item is not returnable. View return policy
216959-92-1
C9H7N3O2S
221.23
MFCD00111662
XDZXQDHRHGIPRN-UHFFFAOYSA-N
4-methyl-2-2-pyrazinyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 2-5-carboxy-4-methyl-1,3-thiazol-2-yl pyrazine, 4-methyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-methyl-2-2-pyrazinyl-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl thiazole-5-carboxylic acid, 5-carboxy-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 5-thiazolecarboxylicacid, 4-methyl-2-2-pyrazinyl, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyrazinyl
2776505
4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
CC1=C(SC(=N1)C1=NC=CN=C1)C(O)=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 221.24
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-(2-pyrazinyl)-1, 3-thiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'