missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(2-pyridyl)thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-7034063
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
34418-48-9
C10H8N2O2S
220.246
MFCD01316364
QFDHWPOPCNNAOI-UHFFFAOYSA-N
4-methyl-2-pyridin-2-yl thiazole-5-carboxylic acid, 2-2-pyridyl-4-methylthiazole-5-carboxylic acid, 4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-2-pyridyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyridyl, 4-methyl-2-pyridin-2-yl-thiazole-5-carboxylic acid, 4-methyl-2-2-pyridinyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-2-pyridyl-1,3-thiazole-5-carboxylic acid, acmc-20ahsp, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyridinyl
214841
4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylic acid
CC1=C(SC(=N1)C2=CC=CC=N2)C(=O)O
This item is not returnable. View return policy
34418-48-9
C10H8N2O2S
220.246
MFCD01316364
QFDHWPOPCNNAOI-UHFFFAOYSA-N
4-methyl-2-pyridin-2-yl thiazole-5-carboxylic acid, 2-2-pyridyl-4-methylthiazole-5-carboxylic acid, 4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-2-pyridyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyridyl, 4-methyl-2-pyridin-2-yl-thiazole-5-carboxylic acid, 4-methyl-2-2-pyridinyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-2-pyridyl-1,3-thiazole-5-carboxylic acid, acmc-20ahsp, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyridinyl
214841
4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylic acid
CC1=C(SC(=N1)C2=CC=CC=N2)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 224°C to 228°C
Quantity 1 g
Formula Weight 220.25
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-(2-pyridyl)thiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'