missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbaldehyde, 97+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carbaldehyde, 4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carbaldehyde, methyltrifluoromethylphenylthiazolecarbaldehyde, 4-methyl-2-4-trifluoromethyl-phenyl-thiazole-5-carbaldehyde, 4-methyl-2-4-trifluoromethylphenyl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carboxaldehyde, 5-thiazolecarboxaldehyde, 4-methyl-2-4-trifluoromethyl phenyl, 5-thiazolecarboxaldehyde,4-methyl-2-4-trifluoromethyl phenyl |
Formula Weight | 271.26 |
Percent Purity | 95% |
Chemical Name or Material | 4-Methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'