Läs mer
4-Methyl-2(5H)-furanone, TRC
Produktkod. 30479701
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30479701 1 g
1 options
Produktkod. 30479701 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3277831G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Methyl-2(5H)-furanone
CAS 6124-79-4
Molekylformel C5 H6 O2
Kvantitet 1 g
Synonym 2(5H)-Furanone, 4-methyl- (8CI, 9CI, ACI), 4-Methyl-2(5H)-furanone (ACI), Crotonic acid, 4-hydroxy-3-methyl-, γ-lactone (6CI, 7CI), 2,5-Dihydro-4-methyl-2-furanone, 3-Methyl-2H-furan-5-one, 3-Methylbut-2-en-4-olide, 3-Methylbut-2-enolide, 4-Hydroxy-3-methyl-2-butenoic acid lactone, 4-Hydroxy-3-methyl-2-butenoic acid γ-lactone, 4-Methyl-2,5-dihydrofuran-2-one, 4-Methyl-5H-furan-2-one
InChI formel InChI=1S/C5H6O2/c1-4-2-5(6)7-3-4/h2H,3H2,1H3
LEDER CC1=CC(=O)OC1
IUPAC-namn 3-methyl-2H-furan-5-one
Molekylvikt (g/mol) 98.1
Formel vikt 98.0368