Läs mer
4-Methyl-2-oxovaleric Acid Sodium Salt Monohydrate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-Methyl-2-oxovaleric Acid Sodium Salt Monohydrate |
| CAS | 313222-86-5 |
| Molekylformel | C6H9NaO3 . H2O |
| Kvantitet | 5 g |
| Synonym | 2-Ketoisocaproic Acid Sodium Salt Monohydrate, 2-Oxo-4-methylpentanoic Acid Sodium Salt Monohydrate, 4-Methyl-2-oxopentanoic Acid Sodium Salt Monohydrate, Sodium 2-Ketoisocaproate Monohydrate, Sodium 4-methyl-2-oxopentanoate Monohydrate, Sodium 4-methyl-2-oxovalerate Monohydrate, Sodium alpha-Keto-isocaproate Monohydrate, Sodium alpha-Ketoisocaproate Monohydrate, Sodium alpha-oxoisocaproate Monohydrate |
| InChI formel | InChI=1S/C6H10O3.Na.H2O/c1-4(2)3-5(7)6(8)9;;/h4H,3H2,1-2H3,(H,8,9);;1H2/q;+1;/p-1 |
| LEDER | O.[Na+].CC(C)CC(=O)C(=O)[O-] |
| IUPAC-namn | sodium;4-methyl-2-oxopentanoate;hydrate |
| Molekylvikt (g/mol) | 170.14 |
| Formel vikt | 170.0555 |