Learn More
4-Methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride, Technical, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 273.76 |
Percent Purity | 95% |
Chemical Name or Material | 4-Methyl-2-phenyl-1, 3-thiazole-5-sulfonyl chloride |