missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole, 97%, Maybridge

946.07 SEK - 8030.85 SEK

Chemical Identifiers

CAS 690632-24-7
Molecular Formula C16H20BNO2S
Molecular Weight (g/mol) 301.211
MDL Number MFCD05865106
InChI Key QJROOQWDMKQWEZ-UHFFFAOYSA-N
Synonym 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-thiazole, 4-methyl-2-phenyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-thiazole, 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiazole, 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1,3-thiazole
PubChem CID 2795497
IUPAC Name 4-methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(S2)C3=CC=CC=C3)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10005722 SDS View Documents Maybridge
CC29239CB
250mg Amber Glass Bottle 946.07 SEK
250mg View alternative products
Save Up to 
»
Estimated Shipment
12-03-2020
Add to basket
 
10695433 SDS View Documents Maybridge
CC29239DA
1g Amber Glass Bottle 2897.20 SEK
1g View alternative products
Save Up to 
»
Estimated Shipment
12-03-2020
Add to basket
 
11316835 SDS View Documents Maybridge
CC29239DE
5g Amber Glass Bottle 8030.85 SEK
5g View alternative products
Save Up to 
»
Estimated Shipment
12-03-2020
Add to basket
 
Specifications

Chemical Identifiers

690632-24-7
301.211
QJROOQWDMKQWEZ-UHFFFAOYSA-N
2795497
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(S2)C3=CC=CC=C3)C
C16H20BNO2S
MFCD05865106
4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-thiazole, 4-methyl-2-phenyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-thiazole, 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiazole, 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1,3-thiazole
4-methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole

Specifications

4-Methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
250mg
96.5
97%
MFCD05865106
QJROOQWDMKQWEZ-UHFFFAOYSA-N
4-methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
2795497
97%
Amber Glass Bottle
690632-24-7
100.0
C16H20BNO2S
4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-thiazole, 4-methyl-2-phenyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-thiazole, 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiazole, 4-methyl-2-phenyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1,3-thiazole
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(S2)C3=CC=CC=C3)C
301.211
301.22