missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
4-Methyl-2-propyl-1H-benzimidazole-6-carboxylic Acid, Mikromol™
Beskrivning
4-Methyl-2-propyl-1H-benzimidazole-6-carboxylic Acid
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1H-Benzimidazole-6-carboxylic-4-methyl-2-propyl |
| CAS | 152628-03-0 |
| Analyt- eller komponentnamn | 1H-Benzimidazole-6-carboxylic-4-methyl-2-propyl |
| Frakt skick | Room Temperature |
| Molekylformel | C12 H14 N2 O2 |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | 2-Propyl-4-methylbenzimidazol-6-carboxylic acid, 4-Methyl-2-propyl-1H-benzimidazole-6-carboxylic acid, 4-Methyl-2-propyl-6-benzimidazolecarboxylic acid, 7-Methyl-2-propyl-3H-benzimidazole-5-carboxylic acid, 1H-Benzimidazole-5-carboxylic acid, 7-methyl-2-propyl-, 1H-Benzimidazole-4-methyl-2-propyl-6-carboxylic Acid |
| InChI formel | InChI=1S/C12H14N2O2/c1-3-4-10-13-9-6-8(12(15)16)5-7(2)11(9)14-10/h5-6H,3-4H2,1-2H3,(H,13,14)(H,15,16) |
| LEDER | CCCc1nc2c(C)cc(cc2[nH]1)C(=O)O |
| IUPAC-namn | 7-methyl-2-propyl-3H-benzimidazole-5-carboxylic acid |
| Visa mer |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?