missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-2-pyrid-4-yl-1,3-thiazol-5-yl)methanol, 97%, Thermo Scientific™

Catalog Number p-3586536
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
886851-57-6
C10H10N2OS
206.26
MFCD09817497
WSBUFMYJALKFHJ-UHFFFAOYSA-N
4-methyl-2-pyridin-4-yl thiazol-5-yl methanol, 4-methyl-2-pyridin-4-yl-1,3-thiazol-5-yl methanol, 4-methyl-2-pyrid-4-yl-1,3-thiazol-5-yl methanol, 5-hydroxymethyl-4-methyl-2-pyridin-4-yl-1,3-thiazole, 5-thiazolemethanol,4-methyl-2-4-pyridinyl, 4-methyl-2-4-pyridyl-1,3-thiazol-5-yl methan-1-ol
20110144
CC1=C(CO)SC(=N1)C1=CC=NC=C1
This item is not returnable. View return policy
886851-57-6
C10H10N2OS
206.26
MFCD09817497
WSBUFMYJALKFHJ-UHFFFAOYSA-N
4-methyl-2-pyridin-4-yl thiazol-5-yl methanol, 4-methyl-2-pyridin-4-yl-1,3-thiazol-5-yl methanol, 4-methyl-2-pyrid-4-yl-1,3-thiazol-5-yl methanol, 5-hydroxymethyl-4-methyl-2-pyridin-4-yl-1,3-thiazole, 5-thiazolemethanol,4-methyl-2-4-pyridinyl, 4-methyl-2-4-pyridyl-1,3-thiazol-5-yl methan-1-ol
20110144
CC1=C(CO)SC(=N1)C1=CC=NC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name [4-methyl-2-(pyridin-4-yl)-1,3-thiazol-5-yl]methanol
Formula Weight 206.27
Percent Purity 97%
Chemical Name or Material (4-Methyl-2-pyrid-4-yl-1, 3-thiazol-5-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'