Läs mer
4-Methyl-3-penten-2-ol, TRC
Produktkod. 30179575
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179575 250 mg
30179542 50 mg
30179368 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179575 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM222478250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4325-82-0
Molekylformel C6H12O
Molekylvikt (g/mol) 100.15
IUPAC-namn 4-methylpent-3-en-2-ol
LEDER CC(O)C=C(C)C
Specifikationer
Kemiskt namn eller material 4-Methyl-3-penten-2-ol
Kvantitet 250 mg
InChI formel InChI=1S/C6H12O/c1-5(2)4-6(3)7/h4,6-7H,1-3H3
Formel vikt 100.0888