missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-(trifluoromethyl)benzoic acid, 97+%, Thermo Scientific™

Catalog Number 11491287
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
261952-01-6
C9H7F3O2
204.15
MFCD01631592
CAPKAYDTKWGFQB-UHFFFAOYSA-N
4-methyl-3-trifluoromethyl benzoic acid, 3-trifluoromethyl-4-methyl benzoic acid, 3-trifluoromethyl-p-toluic acid, mtf-boa-4me, 4-methyl-3-trifluoromethyl benzoicacid, 4-methyl-3-trifluoromethyl-benzoic acid, 3-trifluoromethyl-4-methylbenzoic acid, benzoic acid, 4-methyl-3-trifluoromethyl, pubchem2701
2775592
4-methyl-3-(trifluoromethyl)benzoic acid
CC1=CC=C(C=C1C(F)(F)F)C(O)=O
This item is not returnable. View return policy
261952-01-6
C9H7F3O2
204.15
MFCD01631592
CAPKAYDTKWGFQB-UHFFFAOYSA-N
4-methyl-3-trifluoromethyl benzoic acid, 3-trifluoromethyl-4-methyl benzoic acid, 3-trifluoromethyl-p-toluic acid, mtf-boa-4me, 4-methyl-3-trifluoromethyl benzoicacid, 4-methyl-3-trifluoromethyl-benzoic acid, 3-trifluoromethyl-4-methylbenzoic acid, benzoic acid, 4-methyl-3-trifluoromethyl, pubchem2701
2775592
4-methyl-3-(trifluoromethyl)benzoic acid
CC1=CC=C(C=C1C(F)(F)F)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 175°C to 178°C
Quantity 1 g
Formula Weight 204.15
Percent Purity ≥97%
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'