Läs mer
4-Methyl-5-[(2-aminoethyl)thiomethyl]imidazole Dihydrochloride, TRC
Produktkod. 30472929
Change view
Kvantitet:
200 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472929 200 mg
30476991 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30472929 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM286500200MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38603-72-4
Molekylformel C7 H13 N3 S . 2 Cl H
Molekylvikt (g/mol) 244.19
Synonym Cimetidine Ep Impurity J Dihydrochloride
IUPAC-namn 2-[(4-methyl-1H-imidazol-5-yl)methylsulfanyl]ethanamine;dihydrochloride
LEDER Cl.Cl.Cc1nc[nH]c1CSCCN
Specifikationer
Kemiskt namn eller material 4-Methyl-5-[(2-aminoethyl)thiomethyl]imidazole Dihydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 200 mg
InChI formel InChI=1S/C7H13N3S.2ClH/c1-6-7(10-5-9-6)4-11-3-2-8;;/h5H,2-4,8H2,1H3,(H,9,10);2*1H
Formel vikt 243.0364
Procent renhet >95%