Learn More
Alfa Aesar™ 4-Methyl-5-thiazolemethanol, 97%
Brand:  Alfa Aesar™ H61900.03
Additional Details : CAS Number : 1977-06-6 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15458488
  • 2583.92 SEK / 1g
Estimated Shipment date
from Supplier 03-07-2018
Add to basket
Specifications
Chemical Identifiers
4-Methyl-5-thiazolemethanol
1983-10-4
129.18
(4-Methyl-1,3-thiazol-5-yl)methanol
Specifications
4-Methyl-5-thiazolemethanol
1983-10-4
97%
(4-Methyl-1,3-thiazol-5-yl)methanol
1g
Solid
129.18