Läs mer
4- Methyl Estradiol, TRC
Produktkod. 30137539
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137539 25 mg
30010440 10 mg
30010674 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137539 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30469025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6171-48-8
Molekylformel C19 H26 O2
Molekylvikt (g/mol) 286.41
Synonym 4-Methylestra-1,3,5(10)-triene-3,17beta-diol, Estradiol Hemihydrate Imp. C (EP), 4-Methylestradiol
IUPAC-namn (8R,9S,13S,14S,17S)-4,13-dimethyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
LEDER Cc1c(O)ccc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc12
Specifikationer
Kemiskt namn eller material 4-Methylestradiol
Kvantitet 25 mg
InChI formel InChI=1S/C19H26O2/c1-11-12-3-4-15-14(13(12)5-7-17(11)20)9-10-19(2)16(15)6-8-18(19)21/h5,7,14-16,18,20-21H,3-4,6,8-10H2,1-2H3/t14-,15-,16+,18+,19+/m1/s1
Formel vikt 286.1933