Läs mer
4-Methyl Ethynyl Estradiol, TRC
Produktkod. 30134155
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134155 25 mg
30134146 50 mg
30040897 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134155 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30496525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 155683-61-7
Molekylformel C21 H26 O2
Molekylvikt (g/mol) 310.43
Synonym (17α)-4-Methyl-19-norpregna-1,3,5(10)-trien-20-yne-3,17-diol (9CI), 4-Methyl Ethynyl Estradiol, 4-Methyl-19-nor-17α-pregna-1,3,5(10)-trien-20-yne-3,17-diol (4-Methyl-ethinylestradiol), Ethinylestradiol Imp K. (EP)
IUPAC-namn (8R,9S,13S,14S,17R)-17-ethynyl-4,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
LEDER Cc1c(O)ccc2[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(O)C#C)[C@@H]3CCc12
Specifikationer
Kemiskt namn eller material 4-Methyl Ethynyl Estradiol
Kvantitet 25 mg
InChI formel InChI=1S/C21H26O2/c1-4-21(23)12-10-18-17-6-5-14-13(2)19(22)8-7-15(14)16(17)9-11-20(18,21)3/h1,7-8,16-18,22-23H,5-6,9-12H2,2-3H3/t16-,17-,18+,20+,21+/m1/s1
Formel vikt 310.1933