Läs mer
4-Methyl Phencyclidine Hydrochloride, TRC
Produktkod. 30470484
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470484 5 mg
30477178 1 mg
30478074 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30470484 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3255605MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1934-42-5
Molekylformel C18 H27 N . Cl H
Molekylvikt (g/mol) 293.87
Synonym 4-Me PCP, 4-Me Phencyclidine, 4-methyl Phencyclidine
IUPAC-namn 1-[1-(p-tolyl)cyclohexyl]piperidine;hydrochloride
LEDER Cl.Cc1ccc(cc1)C2(CCCCC2)N3CCCCC3
Specifikationer
Kemiskt namn eller material 4-methyl PCP (hydrochloride)
Kvantitet 5 mg
InChI formel InChI=1S/C18H27N.ClH/c1-16-8-10-17(11-9-16)18(12-4-2-5-13-18)19-14-6-3-7-15-19;/h8-11H,2-7,12-15H2,1H3;1H
Formel vikt 293.191