Läs mer
4-Methylamino Antipyrine, TRC
Produktkod. 17973235
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
17973235 10 mg
1 options
Produktkod. 17973235 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28560010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Methylamino antipyrine
CAS 519-98-2
Anteckningar om renhetsgrad HPLC
Molekylformel C12 H15 N3 O
Kvantitet 10 mg
Synonym 1,2-Dihydro-1,5-dimethyl-4-(methylamino)-2-phenyl-3H-pyrazol-3-one, Antipyrine, 4-(methylamino)- (6CI,8CI), 4-(Methylamino)antipyrine, 4-Methylaminophenazone, 4-Monomethylaminoantipyrine, 4-Monomethylaminophenazone, Methylaminoantipyrine, Methylaminophenazone, Monomethylaminoantipyrine, N-Methyl-4-aminoantipyrine, N-Methyl-4-aminophenazone, N-Methylaminoantipyrine, N-Methylaminophenazone, Noramidopyrine, Noraminopyrine, Metamizole Sodium Monohydrate Imp. C (EP)
InChI formel InChI=1S/C12H15N3O/c1-9-11(13-2)12(16)15(14(9)3)10-7-5-4-6-8-10/h4-8,13H,1-3H3
LEDER CNC1=C(C)N(C)N(C1=O)c2ccccc2
IUPAC-namn 1,5-dimethyl-4-(methylamino)-2-phenylpyrazol-3-one
Molekylvikt (g/mol) 217.27
Formel vikt 217.1215
Procent renhet >95%