Läs mer
4-(Methylamino)cyclohexanone 2,2-Dimethyltrimethylene Ketal Hydrochloride, TRC
Produktkod. 30474957
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30474957 100 mg
1 options
Produktkod. 30474957 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM220805100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(Methylamino)cyclohexanone 2,2-Dimethyltrimethylene Ketal Hydrochloride
CAS 158747-10-5
Molekylformel C12H23NO2.HCl
Kvantitet 100 mg
Synonym 1,5-Dioxaspiro[5.5]undecan-9-amine, N,3,3-trimethyl-, hydrochloride (1:1), 1,5-Dioxaspiro[5.5]undecan-9-amine, N,3,3-trimethyl-, hydrochloride (9CI), 4-Methylaminocyclohexanone (2,2-dimethyltrimethylene) ketal hydrochloride, 4-Methylaminocyclohexanone 2',2'-Dimethyltrimethylene Ketal Hydrochloride, 1,5-Dioxaspiro[5.5]undecan-9-amine, N,3,3-trimethyl-, hydrochloride (1:1), 1,5-Dioxaspiro[5.5]undecan-9-amine, N,3,3-trimethyl-, hydrochloride (9CI), 4-Methylaminocyclohexanone (2,2-dimethyltrimethylene) ketal hydrochloride, 4-Methylaminocyclohexanone 2',2'-Dimethyltrimethylene Ketal Hydrochloride
InChI formel InChI=1S/C12H23NO2.ClH/c1-11(2)8-14-12(15-9-11)6-4-10(13-3)5-7-12;/h10,13H,4-9H2,1-3H3;1H
LEDER Cl.CNC1CCC2(CC1)OCC(C)(C)CO2
IUPAC-namn N,3,3-trimethyl-1,5-dioxaspiro[5.5]undecan-9-amine;hydrochloride
Molekylvikt (g/mol) 249.78
Formel vikt 249.1496