missing translation for 'onlineSavingsMsg'
Learn More

4-Methylanisole, 99%, Thermo Scientific Chemicals

Catalog Number p-3779026
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
104-93-8
C8H10O
122.17
MFCD00008413
CHLICZRVGGXEOD-UHFFFAOYSA-N
4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether
7731
1-methoxy-4-methylbenzene
CC1=CC=C(C=C1)OC
This item is not returnable. View return policy
104-93-8
C8H10O
122.17
MFCD00008413
CHLICZRVGGXEOD-UHFFFAOYSA-N
4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether
7731
1-methoxy-4-methylbenzene
CC1=CC=C(C=C1)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -50°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 174°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4OCH3
Refractive Index 1.5100 to 1.513
Quantity 5 g
Beilstein 06, 392
Specific Gravity 0.96
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alchol
Formula Weight 122.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methylanisole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'