Läs mer
4-Methylbenzotriazole, TRC
Produktkod. 30137657
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137657 500 mg
30133661 1 g
17972505 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137657 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM288780500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29878-31-7
Molekylformel C7 H7 N3
Molekylvikt (g/mol) 133.15
Synonym 4-Methylbenzotriazole, 1H-Benzotriazole, 7-methyl-, 1H-Benzotriazole, 4-methyl- (8CI,9CI), Benzotriazole, 4-methyl- (6CI), 7-Methyl-1H-benzotriazole, 4-Methyl-1,2,3-benzotriazole, 4-Methyl-1H-benzo[d][1,2,3]triazole, 4-Methyl-1H-benzotriazole, 4-Methylbenzotriazole, 1H-Benzotriazole, 7-methyl-, 1H-Benzotriazole, 4-methyl- (8CI,9CI), Benzotriazole, 4-methyl- (6CI), 7-Methyl-1H-benzotriazole, 4-Methyl-1,2,3-benzotriazole, 4-Methyl-1H-benzo[d][1,2,3]triazole, 4-Methyl-1H-benzotriazole
IUPAC-namn 4-methyl-1H-benzotriazole
LEDER Cc1cccc2[nH]nnc12
Specifikationer
Kemiskt namn eller material 4-Methylbenzotriazole
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C7H7N3/c1-5-3-2-4-6-7(5)9-10-8-6/h2-4H,1H3,(H,8,9,10)
Formel vikt 133.064
Procent renhet >95%