missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-7021438
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
104-84-7
C8H11N
121.183
MFCD00008123
HMTSWYPNXFHGEP-UHFFFAOYSA-N
4-methylbenzylamine, benzenemethanamine, 4-methyl, p-methylbenzylamine, p-tolylmethanamine, 4-methylphenyl methanamine, benzylamine, p-methyl, p-xylylamine, 4-methybenzylamine, 1-4-methylphenyl methanamine, 4-methyl-benzylamine
66035
(4-methylphenyl)methanamine
CC1=CC=C(C=C1)CN
This item is not returnable. View return policy
104-84-7
C8H11N
121.183
MFCD00008123
HMTSWYPNXFHGEP-UHFFFAOYSA-N
4-methylbenzylamine, benzenemethanamine, 4-methyl, p-methylbenzylamine, p-tolylmethanamine, 4-methylphenyl methanamine, benzylamine, p-methyl, p-xylylamine, 4-methybenzylamine, 1-4-methylphenyl methanamine, 4-methyl-benzylamine
66035
(4-methylphenyl)methanamine
CC1=CC=C(C=C1)CN

The reactions of 1-hexanol with benzylamine and with 4-methylbenzylamine led to the corresponding imines in moderate yields. 4-methylbenzylamine in ethanol/water solution (1:4) as simultaneously absorbing and buffering background electrolyte with detection at 210 nm was found suitable for determination of the individual compounds in determination by capillary zone electrophoresis with indirect detection. Synthesis of benzimidazoles uses 4-methylbenzylamine (R 1 =4-MeBn) and 4-chlorophenyl isothiocyanate (R 2 =4-ClPh) as building blocks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The reactions of 1-hexanol with benzylamine and with 4-methylbenzylamine led to the corresponding imines in moderate yields. 4-methylbenzylamine in ethanol/water solution (1:4) as simultaneously absorbing and buffering background electrolyte with detection at 210 nm was found suitable for determination of the individual compounds in determination by capillary zone electrophoresis with indirect detection. Synthesis of benzimidazoles uses 4-methylbenzylamine (R 1 =4-MeBn) and 4-chlorophenyl isothiocyanate (R 2 =4-ClPh) as building blocks.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 12°C to 13°C
Density 0.955
Boiling Point 196°C to 197°C
Flash Point 75°C (167°F)
Refractive Index 1.535
Quantity 25 g
UN Number UN2735
Beilstein 956670
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 121.18
Percent Purity 98%
Chemical Name or Material 4-Methylbenzylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'