missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylbiphenyl-4-sulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7033233
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
101366-51-2
C13H11ClO2S
266.739
MFCD01631922
DBYXACYHGMSPMQ-UHFFFAOYSA-N
4'-methylbiphenyl-4-sulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methylphenyl benzenesulfonyl chloride, 4-4-methylphenyl phenyl sulphonyl chloride, 4'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 4-p-tolyl benzenesulfonyl chloride, acmc-20amef, 4-4-chlorosulphonyl phenyl toluene, 4-chlorosulphonyl-4'-methylbiphenyl
2794758
4-(4-methylphenyl)benzenesulfonyl chloride
CC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
This item is not returnable. View return policy
101366-51-2
C13H11ClO2S
266.739
MFCD01631922
DBYXACYHGMSPMQ-UHFFFAOYSA-N
4'-methylbiphenyl-4-sulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methylphenyl benzenesulfonyl chloride, 4-4-methylphenyl phenyl sulphonyl chloride, 4'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 4-p-tolyl benzenesulfonyl chloride, acmc-20amef, 4-4-chlorosulphonyl phenyl toluene, 4-chlorosulphonyl-4'-methylbiphenyl
2794758
4-(4-methylphenyl)benzenesulfonyl chloride
CC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 266.74
Percent Purity 98%
Chemical Name or Material 4'-Methylbiphenyl-4-sulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'