missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanemethanol, TRC

Product Code. 30476797
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
2.5 g
missing translation for 'unitSize'
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30476797 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30476797 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM2949152.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-Methylcyclohexanemethanol
CAS 34885-03-5
Molekylformel C8H16O
Kvantitet 2.5 g
Synonym 4-Methyl-cyclohexanemethanol, (4-Methylcyclohexyl)methanol, 4-Methylcyclohexane methanol, Hexahydro-p-methylbenzyl alcohol
InChI formel InChI=1S/C8H16O/c1-7-2-4-8(6-9)5-3-7/h7-9H,2-6H2,1H3
LEDER CC1CCC(CO)CC1
IUPAC-namn (4-methylcyclohexyl)methanol
Molekylvikt (g/mol) 128.21
Formel vikt 128.1201
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.