Läs mer
(4-Methylene-5-hexenyl)triphenyl-phosphonium Iodide, TRC
Produktkod. 30477350
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477350 10 mg
30481094 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30477350 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30410510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4-Methylene-5-hexenyl)triphenyl-phosphonium Iodide
CAS 17842-92-1
Molekylformel C25H26IP
Kvantitet 10 mg
Synonym (4-Methylene-5-hexen-1-yl)triphenyl-phosphonium Iodide
InChI formel InChI=1S/C25H26P.HI/c1-3-22(2)14-13-21-26(23-15-7-4-8-16-23,24-17-9-5-10-18-24)25-19-11-6-12-20-25;/h3-12,15-20H,1-2,13-14,21H2;1H/q+1;/p-1
LEDER [I-].C=CC(=C)CCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC-namn 4-methylidenehex-5-enyl(triphenyl)phosphanium;iodide
Molekylvikt (g/mol) 484.35
Formel vikt 484.0817