Läs mer
4-Methylheptane, TRC
Produktkod. 30481555
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481555 500 mg
30481554 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30481555 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM312430500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Methylheptane
CAS 589-53-7
Molekylformel C8 H18
Kvantitet 500 mg
Synonym Heptane','4-methyl-, 4-Methylheptane, NSC 73953
InChI formel InChI=1S/C8H18/c1-4-6-8(3)7-5-2/h8H,4-7H2,1-3H3
LEDER CCCC(C)CCC
IUPAC-namn 4-methylheptane
Molekylvikt (g/mol) 114.23
Formel vikt 114.1409