Läs mer
Tocris Bioscience™ 4-Methylhistamine dihydrochloride
Selective, high affinity H4 agonist
Brand: Tocris Bioscience™ 2342/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
4-Methylhistamine dihydrochloride is a potent, high affinity H4 receptor agonist (Ki = 7 nM) that displays > 100-fold selectivity over other human histamine receptor subtypes.Specifikationer
| 5-(2-Aminoethyl)-4-methylimidazole dihydrochloride | |
| 36376-47-3 | |
| 10 mg | |
| Soluble to 50mM in water |
Produkttitel
For Research Use Only