missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylmorpholine N-oxide monohydrate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
Description
4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in a cool place. Keep the container tightly closed. Incompatible with strong oxidizing agents.
4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in a cool place. Keep the container tightly closed. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 73°C to 76°C |
Quantity | 10 g |
Beilstein | 5449441 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 135.17 (117.15 Anhydrous) |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Methylmorpholine N-oxide monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'