Läs mer
4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol N-Beta-D-Glucuronide, TRC
Produktkod. 17951425
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17951425 1 mg
17933035 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17951425 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3257451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 350508-29-1
Molekylformel C16 H23 N3 O8
Molekylvikt (g/mol) 385.37
Synonym Pyridinium, 1-β-D-glucopyranuronosyl-3-[(1S)-1-hydroxy-4-(methylnitrosoamino)butyl]-, inner salt, stereoisomer (9CI), 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol N-β-D-glucuronide, NNAL-N-glucuronide
IUPAC-namn (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[4-[methyl(nitroso)amino]-1-pyridin-3-ylbutoxy]oxane-2-carboxylic acid
LEDER CN(CCCC(O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O)c2cccnc2)N=O
Specifikationer
Kemiskt namn eller material 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol N-beta-D-Glucuronide
Kvantitet 1 mg
InChI formel InChI=1S/C16H23N3O8/c1-19(18-25)7-3-5-10(9-4-2-6-17-8-9)26-16-13(22)11(20)12(21)14(27-16)15(23)24/h2,4,6,8,10-14,16,20-22H,3,5,7H2,1H3,(H,23,24)/t10?,11-,12-,13+,14-,16+/m0/s1
Formel vikt 385.1485