Läs mer
4-(Methylnitrosamino)-1-(3-pyridyl-N-oxide)-1-butanol, TRC
Produktkod. 30136071
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136071 50 mg
30008113 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136071 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32575550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 85352-99-4
Molekylformel C10 H15 N3 O3
Molekylvikt (g/mol) 225.24
Synonym 3-Pyridinemethanol, α-[3-(methylnitrosoamino)propyl]-, 1-oxide, 4-(Methylnitrosamino)-1-(3-pyridyl-N-oxide)-1-butanol, α-[3-(Methylnitrosoamino)propyl]-3-pyridinemethanol 1-oxide, 4-(N-Methyl-N-nitrosamino)-1-(3-pyridyl-N-oxide)butan-1-ol
IUPAC-namn N-[4-hydroxy-4-(1-oxidopyridin-1-ium-3-yl)butyl]-N-methylnitrous amide
LEDER CN(CCCC(O)c1ccc[n+]([O-])c1)N=O
Specifikationer
Kemiskt namn eller material 4-(Methylnitrosamino)-1-(3-pyridyl-N-oxide)-1-butanol
Kvantitet 50 mg
InChI formel InChI=1S/C10H15N3O3/c1-12(11-15)6-3-5-10(14)9-4-2-7-13(16)8-9/h2,4,7-8,10,14H,3,5-6H2,1H3
Formel vikt 225.1113