missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylpentan-2-One, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Specifications
Melting Point | -84°C |
Color | Colorless |
Boiling Point | 117.4°C |
Packaging | Amber glass bottle |
Quantity | 1 L |
Formula Weight | 100.16g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- 4-Methylpentan-2-one
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'