Läs mer
4-Methylpentanal (up to 75% hydrated form), TRC
Produktkod. 30135049
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135049 500 mg
30134618 100 mg
17993785 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135049 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM325550500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1119-16-0
Molekylformel C6H12O
Molekylvikt (g/mol) 100.16
Synonym 4-Methylvaleraldehyde, 4-Methyl-1-pentanal, 4-Methylpentan-1-one, Isocaproaldehyde, Isohexanal, γ-Methylvaleraldehyde;
IUPAC-namn 4-methylpentanal
LEDER CC(C)CCC=O
Specifikationer
Kemiskt namn eller material 4-Methylpentanal
Kvantitet 500 mg
InChI formel InChI=1S/C6H12O/c1-6(2)4-3-5-7/h5-6H,3-4H2,1-2H3
Formel vikt 100.0888